Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrnb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_052806.2 | NP_434693.1 | 2 - 3 | 193 | 85 | |
GenBank mRNA | AY574260.1 | - | 2 - 3 | 193 | 85 | |
U42976.1 | - | 253 | 85 | |||
X15834.1 | - | 305 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Rattus norvegicus Probe spans exons |