Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109292.1 | NP_001102762.1 | 6 - 7 | 996 | 121 | |
XM_006242950.2 | XP_006243012.1 | 6 - 7 | 996 | 121 | ||
XM_006242951.2 | XP_006243013.1 | 6 - 7 | 1010 | 121 | ||
XM_006242952.2 | XP_006243014.1 | 6 - 7 | 1005 | 121 | ||
XM_006242954.2 | XP_006243016.1 | 6 - 7 | 1279 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |