Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Peak1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108149.1 | NP_001101619.1 | 3 - 4 | 344 | 105 | |
XM_006243115.2 | XP_006243177.1 | 2 - 3 | 214 | 105 | ||
XM_006243116.2 | XP_006243178.1 | 4 - 5 | 966 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |