Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Paqr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014092.1 | NP_001014114.1 | 5 - 6 | 833 | 90 | |
XM_006243208.2 | XP_006243270.1 | 5 - 6 | 765 | 90 | ||
XM_006243209.2 | XP_006243271.1 | 4 - 5 | 1102 | 90 | ||
XM_008766275.1 | XP_008764497.1 | 5 - 6 | 693 | 90 | ||
XM_008766276.1 | XP_008764498.1 | 4 - 5 | 637 | 90 | ||
GenBank mRNA | BC087040.1 | - | 5 - 6 | 833 | 7106223 | 90 |
DQ088965.1 | - | 4 - 5 | 507 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |