Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yes1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033298.1 | NP_150640.1 | 7 - 8 | 1094 | 92 | |
XM_006245675.2 | XP_006245737.1 | 7 - 8 | 1107 | 92 | ||
GenBank mRNA | AB037472.1 | - | 6 - 7 | 871 | 92 | |
BC079403.1 | - | 7 - 8 | 1094 | 7112783 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |