Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdac8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126373.2 | NP_001119845.2 | 5 - 6 | 887 | 91 | |
XM_006257123.2 | XP_006257185.1 | 5 - 6 | 895 | 91 | ||
XM_006257124.2 | XP_006257186.1 | 5 - 6 | 894 | 91 | ||
GenBank mRNA | BC162023.1 | - | 5 - 6 | 880 | 9099275 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Rattus norvegicus Probe spans exons |