Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itgb1bp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108245.2 | NP_001101715.1 | 3 - 4 | 226 | 83 | |
GenBank mRNA | FM116382.1 | - | 3 - 4 | 185 | 83 | |
FM118664.1 | - | 3 - 4 | 210 | 83 | ||
FQ216044.1 | - | 3 - 4 | 225 | 83 | ||
FQ216100.1 | - | 3 - 4 | 202 | 83 | ||
FQ224925.1 | - | 3 - 4 | 223 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |