Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ano8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006252965.2 | XP_006253027.2 | 11 - 12 | 1498 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |