Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019313.1 | NP_062186.1 | 6 - 7 | 1289 | 74 | |
GenBank mRNA | AF000973.1 | - | 6 - 7 | 1289 | 74 | |
U69885.1 | - | 6 - 7 | 1055 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Rattus norvegicus Probe spans exons |