Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem59l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271055.1 | NP_001257984.1 | 1 - 2 | 204 | 78 | |
GenBank mRNA | BC089056.1 | - | 1 - 2 | 146 | 78 | |
FQ212329.1 | - | 1 - 2 | 178 | 78 | ||
FQ214027.1 | - | 1 - 2 | 158 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |