Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem59l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271055.1 | NP_001257984.1 | 2 - 3 | 354 | 62 | |
XM_006252897.2 | XP_006252959.1 | 3 - 4 | 424 | 62 | ||
GenBank mRNA | BC089056.1 | - | 2 - 3 | 296 | 62 | |
FQ212329.1 | - | 2 - 3 | 328 | 62 | ||
FQ214027.1 | - | 2 - 3 | 308 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |