Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cope | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006252876.2 | XP_006252938.1 | 8 - 9 | 1022 | 84 | |
GenBank mRNA | BC161928.1 | - | 6 - 7 | 2021 | 8374450 | 84 |
FQ220757.1 | - | 8 - 9 | 832 | 84 | ||
FQ232202.1 | - | 8 - 9 | 831 | 84 | ||
FQ233163.1 | - | 8 - 9 | 832 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons |