Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifi30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030026.1 | NP_001025197.1 | 3 - 4 | 510 | 95 | |
GenBank mRNA | BC088256.1 | - | 3 - 4 | 374 | 95 | |
BC099774.1 | - | 3 - 4 | 510 | 7932222 | 95 | |
FQ225096.1 | - | 3 - 4 | 396 | 95 | ||
FQ229307.1 | - | 3 - 4 | 548 | 95 | ||
FQ230297.1 | - | 3 - 4 | 396 | 95 | ||
FQ233058.1 | - | 3 - 4 | 396 | 95 | ||
FQ233063.1 | - | 3 - 4 | 428 | 95 | ||
FQ233214.1 | - | 3 - 4 | 396 | 95 | ||
FQ234315.1 | - | 3 - 4 | 396 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |