Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rfxank | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013136.1 | NP_001013154.1 | 4 - 5 | 786 | 65 | |
XM_006252898.2 | XP_006252960.1 | 5 - 6 | 660 | 65 | ||
XM_006252899.2 | XP_006252961.1 | 5 - 6 | 697 | 65 | ||
XM_006252900.2 | XP_006252962.1 | 4 - 5 | 701 | 65 | ||
GenBank mRNA | BC087111.1 | - | 4 - 5 | 786 | 7193515 | 65 |
FQ231410.1 | - | 4 - 5 | 595 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |