Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rfxank | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013136.1 | NP_001013154.1 | 5 - 6 | 887 | 84 | |
XM_006252898.2 | XP_006252960.1 | 6 - 7 | 761 | 84 | ||
XM_006252899.2 | XP_006252961.1 | 6 - 7 | 798 | 84 | ||
XM_006252900.2 | XP_006252962.1 | 5 - 6 | 802 | 84 | ||
GenBank mRNA | BC087111.1 | - | 5 - 6 | 887 | 7193515 | 84 |
FQ231410.1 | - | 5 - 6 | 696 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |