Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Calr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012212.1 | NP_001012212.1 | 4 - 5 | 676 | 154 | |
GenBank mRNA | BC079049.1 | - | 4 - 5 | 676 | 7115321 | 154 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |