Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cilp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107307.1 | NP_001100777.1 | 2 - 3 | 163 | 153 | |
XM_006252902.2 | XP_006252964.1 | 2 - 3 | 250 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |