Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sftpa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270645.1 | NP_001257574.1 | 4 - 5 | 436 | 87 | |
NM_001270647.1 | NP_001257576.1 | 4 - 5 | 647 | 87 | ||
NM_017329.2 | NP_059025.2 | 4 - 5 | 433 | 87 | ||
GenBank mRNA | BC085353.1 | - | 4 - 5 | 436 | 7121530 | 87 |
CK364490.1 | - | 4 - 5 | 659 | 87 | ||
M15754.1 | - | 4 - 5 | 421 | 87 | ||
M33201.1 | - | 4 - 5 | 430 | 87 | ||
X13176.1 | - | 4 - 5 | 421 | 87 | ||
X13177.1 | - | 4 - 5 | 430 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |