Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcyox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145085.2 | NP_659553.1 | 3 - 4 | 530 | 114 | |
GenBank mRNA | AF332142.1 | - | 520 | 114 | ||
BC078719.1 | - | 3 - 4 | 530 | 7127920 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |