Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cidec | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024333.2 | NP_001019504.2 | 4 - 5 | 416 | 64 | |
NM_001244797.1 | NP_001231726.1 | 4 - 5 | 428 | 64 | ||
NM_001244798.1 | NP_001231727.1 | 4 - 5 | 417 | 64 | ||
GenBank mRNA | BC083860.1 | - | 4 - 5 | 416 | 7126532 | 64 |
FM043477.1 | - | 4 - 5 | 425 | 64 | ||
FM103335.1 | - | 4 - 5 | 417 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |