Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mxd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100749.1 | NP_001094219.1 | 2 - 3 | 301 | 69 | |
GenBank mRNA | AY328517.1 | - | 1 - 2 | 80 | 69 | |
AY386318.1 | - | 2 - 3 | 143 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |