Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crbn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015003.2 | NP_001015003.1 | 5 - 6 | 710 | 148 | |
GenBank mRNA | AY973953.1 | - | 5 - 6 | 697 | 148 | |
BC099779.1 | - | 5 - 6 | 695 | 148 | ||
BC127455.1 | - | 5 - 6 | 702 | 7112865 | 148 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Rattus norvegicus Probe spans exons |