Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Reg1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012641.1 | NP_036773.1 | 3 - 4 | 254 | 103 | |
GenBank mRNA | M18962.1 | - | 2 - 3 | 230 | 103 | |
M62930.1 | - | 3 - 4 | 254 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |