Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nptn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006243172.2 | XP_006243234.1 | 7 - 8 | 1284 | 94 | |
XM_006243174.2 | XP_006243236.1 | 6 - 7 | 945 | 94 | ||
GenBank mRNA | FQ226464.1 | - | 2 - 3 | 144 | 94 | |
X99337.1 | - | 6 - 7 | 974 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |