Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005547.1 | NP_001005547.1 | 4 - 5 | 536 | 124 | |
GenBank mRNA | BC082102.1 | - | 4 - 5 | 536 | 7134588 | 124 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |