Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sdhd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198788.2 | NP_942083.1 | 2 - 3 | 176 | 85 | |
GenBank mRNA | BC059160.1 | - | 2 - 3 | 181 | 6887444 | 85 |
FQ209438.1 | - | 2 - 3 | 177 | 85 | ||
FQ212791.1 | - | 2 - 3 | 179 | 85 | ||
FQ212873.1 | - | 2 - 3 | 179 | 85 | ||
FQ212985.1 | - | 2 - 3 | 178 | 85 | ||
FQ213509.1 | - | 2 - 3 | 178 | 85 | ||
FQ214315.1 | - | 2 - 3 | 206 | 85 | ||
FQ215255.1 | - | 2 - 3 | 183 | 85 | ||
FQ215355.1 | - | 2 - 3 | 177 | 85 | ||
FQ215505.1 | - | 2 - 3 | 178 | 85 | ||
FQ215951.1 | - | 2 - 3 | 178 | 85 | ||
FQ215986.1 | - | 2 - 3 | 177 | 85 | ||
FQ216027.1 | - | 2 - 3 | 178 | 85 | ||
FQ216314.1 | - | 2 - 3 | 184 | 85 | ||
FQ216503.1 | - | 2 - 3 | 181 | 85 | ||
FQ220862.1 | - | 2 - 3 | 177 | 85 | ||
FQ230166.1 | - | 2 - 3 | 170 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |