Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpinb11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107167.1 | NP_001100637.1 | 3 - 4 | 421 | 145 | |
XM_006249632.2 | XP_006249694.2 | 2 - 3 | 571 | 145 | ||
XM_006249633.2 | XP_006249695.2 | 2 - 3 | 273 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |