Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134360.1 | NP_599187.1 | 7 - 8 | 672 | 81 | |
XM_008762465.1 | XP_008760687.1 | 6 - 7 | 672 | 81 | ||
GenBank mRNA | BC097949.1 | - | 7 - 8 | 672 | 7387440 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |