Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnttip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134400.2 | NP_599227.2 | 2 - 3 | 228 | 101 | |
XM_006235512.2 | XP_006235574.1 | 2 - 3 | 247 | 101 | ||
XM_006235513.2 | XP_006235575.1 | 2 - 3 | 196 | 101 | ||
GenBank mRNA | AF348701.1 | - | 2 - 3 | 197 | 101 | |
BC100640.1 | - | 2 - 3 | 228 | 7929876 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |