Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sept12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100865.1 | NP_001094335.1 | 7 - 8 | 796 | 122 | |
GenBank mRNA | BC097401.1 | - | 7 - 8 | 796 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |