Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cluap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014225.1 | NP_001014247.1 | 4 - 5 | 742 | 81 | |
XM_006245832.2 | XP_006245894.1 | 4 - 5 | 505 | 81 | ||
XM_008767512.1 | XP_008765734.1 | 4 - 5 | 438 | 81 | ||
XM_008767513.1 | XP_008765735.1 | 4 - 5 | 800 | 81 | ||
XM_008767516.1 | XP_008765738.1 | 3 - 4 | 344 | 81 | ||
XM_008767517.1 | XP_008765739.1 | 2 - 3 | 207 | 81 | ||
XM_008767518.1 | XP_008765740.1 | 2 - 3 | 264 | 81 | ||
GenBank mRNA | BC079022.1 | - | 4 - 5 | 742 | 7113868 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |