Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tprkb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013926.2 | NP_001013948.2 | 2 - 3 | 261 | 149 | |
XM_003749789.3 | XP_003749837.1 | 2 - 3 | 212 | 149 | ||
XM_006236804.2 | XP_006236866.1 | 3 - 4 | 918 | 149 | ||
XM_006236805.2 | XP_006236867.1 | 3 - 4 | 413 | 149 | ||
XM_006236808.2 | XP_006236870.1 | 3 - 4 | 283 | 149 | ||
XM_008763062.1 | XP_008761284.1 | 4 - 5 | 955 | 149 | ||
GenBank mRNA | BC087060.1 | - | 3 - 4 | 276 | 7125830 | 149 |
CK475733.1 | - | 2 - 3 | 261 | 149 | ||
CV112449.1 | - | 2 - 3 | 189 | 149 | ||
FQ234910.1 | - | 2 - 3 | 218 | 149 |
Gene Symbol: LOC103690067 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006236742.2 | XP_006236804.1 | 918 | 149 | ||
XM_006236743.2 | XP_006236805.1 | 413 | 149 | |||
XM_006236745.2 | XP_006236807.1 | 283 | 149 | |||
XM_006236746.2 | XP_006236808.1 | 197 | 149 | |||
XM_008763108.1 | XP_008761330.1 | 297 | 149 | |||
XM_008763109.1 | XP_008761331.1 | 955 | 149 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |