Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp2b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012508.5 | NP_036640.1 | 131 | 60 | ||
GenBank mRNA | J03754.1 | - | 137 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Transporter Rattus norvegicus Probe spans exons |