Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc6a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024371.2 | NP_077347.1 | 4 - 5 | 622 | 86 | |
XM_006237059.1 | XP_006237121.1 | 5 - 6 | 831 | 86 | ||
XM_006237060.2 | XP_006237122.1 | 5 - 6 | 3051 | 86 | ||
GenBank mRNA | M59742.1 | - | 4 - 5 | 622 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Rattus norvegicus Probe spans exons |