Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053534.2 | NP_445986.2 | 1 - 2 | 107 | 82 | |
GenBank mRNA | FQ228747.1 | - | 1 - 2 | 182 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |