Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elmod3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013087.1 | NP_001013105.1 | 5 - 6 | 543 | 93 | |
XM_006236667.2 | XP_006236729.1 | 5 - 6 | 543 | 93 | ||
XM_006236669.1 | XP_006236731.1 | 4 - 5 | 495 | 93 | ||
XM_008762978.1 | XP_008761200.1 | 4 - 5 | 505 | 93 | ||
GenBank mRNA | BC083623.1 | - | 5 - 6 | 543 | 7134106 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |