Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp4r2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106613.2 | NP_001100083.2 | 6 - 7 | 706 | 124 | |
XM_006236962.2 | XP_006237024.1 | 6 - 7 | 780 | 124 | ||
GenBank mRNA | BC167050.1 | - | 6 - 7 | 624 | 7114402 | 124 |
EV776660.1 | - | 3 - 4 | 185 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |