Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Capg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013086.1 | NP_001013104.1 | 3 - 4 | 499 | 179 | |
XM_006236661.2 | XP_006236723.1 | 4 - 5 | 339 | 179 | ||
XM_006236662.2 | XP_006236724.1 | 4 - 5 | 432 | 179 | ||
XM_006236663.2 | XP_006236725.1 | 4 - 5 | 566 | 179 | ||
XM_006236666.2 | XP_006236728.1 | 3 - 4 | 286 | 179 | ||
GenBank mRNA | BC079104.1 | - | 3 - 4 | 499 | 7125038 | 179 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |