Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Reg3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053289.1 | NP_445741.1 | 1 - 2 | 47 | 110 | |
GenBank mRNA | FQ232415.1 | - | 1 - 2 | 47 | 110 | |
FQ233015.1 | - | 1 - 2 | 47 | 110 | ||
S43715.1 | - | 1 - 2 | 47 | 110 | ||
S77413.1 | - | 1 - 2 | 45 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |