Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpinb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021696.2 | NP_067728.1 | 3 - 4 | 446 | 142 | |
XM_006249639.2 | XP_006249701.1 | 4 - 5 | 632 | 142 | ||
XM_008769495.1 | XP_008767717.1 | 4 - 5 | 529 | 142 | ||
GenBank mRNA | X64563.1 | - | 3 - 4 | 447 | 142 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Detect Genomic DNA Rattus norvegicus Probe spans exons |