Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135715.1 | NP_001129187.1 | 7 - 8 | 2231 | 110 | |
XM_006255815.2 | XP_006255877.1 | 7 - 8 | 2522 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |