Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxw9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081634.1 | NP_001075103.1 | 2 - 3 | 619 | 64 | |
GenBank mRNA | BC084706.1 | - | 2 - 3 | 580 | 64 | |
BC166590.1 | - | 2 - 3 | 554 | 7938435 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |