Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108230.1 | NP_001101700.1 | 2 - 3 | 149 | 104 | |
XM_006245400.2 | XP_006245462.2 | 3 - 4 | 4244 | 104 | ||
XM_006245403.2 | XP_006245465.2 | 3 - 4 | 4244 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |