Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: D2hgdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106926.1 | NP_001100396.1 | 7 - 8 | 1386 | 124 | |
XM_006245520.2 | XP_006245582.1 | 7 - 8 | 1302 | 124 | ||
XM_006245521.2 | XP_006245583.1 | 7 - 8 | 1160 | 124 | ||
XM_006245522.2 | XP_006245584.1 | 7 - 8 | 1155 | 124 | ||
XM_008767365.1 | XP_008765587.1 | 7 - 8 | 1417 | 124 | ||
GenBank mRNA | BC166559.1 | - | 6 - 7 | 1059 | 7312233 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |