Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slco6c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173338.1 | NP_775460.1 | 1877 | 64 | ||
XM_008767361.1 | XP_008765583.1 | 1759 | 64 | |||
GenBank mRNA | AF326113.1 | - | 1877 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Rattus norvegicus Probe spans exons |