Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025012.1 | NP_001020183.1 | 6 - 7 | 1191 | 110 | |
XM_006245604.1 | XP_006245666.1 | 5 - 6 | 1114 | 110 | ||
XM_006245605.2 | XP_006245667.1 | 6 - 7 | 1610 | 110 | ||
XM_008767377.1 | XP_008765599.1 | 6 - 7 | 4786 | 110 | ||
XM_008767378.1 | XP_008765600.1 | 6 - 7 | 4446 | 110 | ||
GenBank mRNA | BC082058.1 | - | 6 - 7 | 1191 | 7132420 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |