Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022666.1 | NP_073157.1 | 1725 | 91 | ||
GenBank mRNA | M90518.1 | - | 1725 | 91 | ||
M92077.1 | - | 940 | 91 | |||
U47331.1 | - | 1725 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Rattus norvegicus Probe spans exons |