Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jmjd1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191719.1 | NP_001178648.1 | 20 - 21 | 6781 | 98 | |
XM_006256359.2 | XP_006256421.1 | 20 - 21 | 6835 | 98 | ||
XM_006256360.2 | XP_006256422.1 | 19 - 20 | 6631 | 98 | ||
XM_006256361.2 | XP_006256423.1 | 20 - 21 | 6858 | 98 | ||
XM_006256362.2 | XP_006256424.1 | 20 - 21 | 6750 | 98 | ||
XM_006256363.1 | XP_006256425.1 | 16 - 17 | 6192 | 98 | ||
XM_006256364.1 | XP_006256426.1 | 20 - 21 | 6744 | 98 | ||
XM_006256365.1 | XP_006256427.1 | 15 - 16 | 6050 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |