Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kifc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005878.1 | NP_001005878.1 | 6 - 7 | 867 | 69 | |
GenBank mRNA | BC083827.1 | - | 6 - 7 | 867 | 7112250 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |