Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC103694871 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008773820.1 | XP_008772042.1 | 1 - 2 | 394 | 85 |
Gene Symbol: Vpreb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006256322.1 | XP_006256384.1 | 1 - 2 | 394 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Rattus norvegicus Probe spans exons |